D-(-)-Sorbitol 98%
Supplier: Thermo Scientific Chemicals
36404-36
36404-A3
CAAA36404-36EA
59.09
CAD
CAAA36404-36
CAAA36404-A3
D-(-)-Sorbitol 98%
D(-)-Sorbitol
MDL: MFCD00004708
Soluble in water, methanol, acetone, DMF
Purity: 98%
CAS No.: 50-70-4
Molecular Formula: C6H14O6
Formula Weight: 182.17
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 98-100°
Solubility: Soluble in water, methanol, acetone, DMF
Beilstein Reference: 1721899
Merck Reference: 14,8725
MDL No.: MFCD00004708
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 295 °C (1013 hPa) Melting Pt: 98…100 °C Density: 1.524 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004708 CAS Number: 50-70-4 EINECS: 200-061-5 Merck Index: 14,8725 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...