Magnesium fluoride ≥99.9% (metals basis excluding Ca, Na) (max. 1% Ca + Na)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36391-14
36391-22
CAAA36391-14EA
121.34
CAD
CAAA36391-14
CAAA36391-22
Magnesium fluoride ≥99.9% (metals basis excluding Ca, Na) (max. 1% Ca + Na)
Magnesium fluoride
Vacuum deposition
Purity: 99.9%
CAS No.: 7783-40-6
Molecular Formula: F2Mg
Formula Weight: 62.30
Physical Form: 3-6mm Vacuum Melted Lump
Density: 3.148
Refractive Index: 1.35-1.38
Appearance: White
Odor: Odorless
Melting Point: 1261°
Merck Reference: 14,5665
MDL No.: MFCD00011108
Vacuum deposition
Formula:
MgF₂ MW: 62.3 g/mol Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1261 °C Density: 3.148 Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...