Iron(III) oxide ≥99.85% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36387-18
36387-30
CAAA36387-18EA
213.3
CAD
CAAA36387-18
CAAA36387-30
Iron(III) oxide ≥99.85% (metals basis)
Iron (III) oxide
MDL: MFCD00011008
Purity: 99.85+%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: 3-12mm Sintered Lump
Density: 5.24
Appearance: Red-brown
Odor: Odorless
Melting Point: 1565°
Merck Reference: 14,4028
MDL No.: MFCD00011008
Formula:
Fe₂O₃ Melting Pt: 1565 °C Density: 5.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...