Indium (III) oxide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Indium trioxide, Diindium trioxide

36386-09 36386-18
CAAA36386-09EA 155.16 CAD
CAAA36386-09 CAAA36386-18
Indium (III) oxide ≥99.999% (metals basis)
Indium (III) oxide

MDL: MFCD00011060 Notes: For vacuum deposition


Purity: 99.999% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: 3-6mm Sintered Granules Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Formula: In₂O₃
MW: 277.64 g/mol
Melting Pt: 715.05
Density: 7.18
Storage Temperature: Ambient
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR