Germanium(IV) oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36382-04
36382-09
36382-18
CAAA36382-04EA
103.33
CAD
CAAA36382-04
CAAA36382-09
CAAA36382-18
Germanium(IV) oxide ≥99.99% (metals basis)
Germanium (IV) oxide
Used in phosphors, transistors and diodes, and infrared transmitting glass
Purity: 99.99%
CAS No.: 1310-53-8
Molecular Formula: GeO2
Formula Weight: 104.59
Physical Form: 3-12mm Sintered Lump
Density: 6.239
Appearance: White
Odor: Odorless
Melting Point: 1086°
Merck Reference: 14,4409
MDL No.: MFCD00011030
Used in phosphors, transistors and diodes, and infrared transmitting glass
Formula:
GeO₂ Melting Pt: 1086 °C Density: 6.239 Storage Temperature: Ambient |
MDL Number:
MFCD00011030 CAS Number: 1310-53-8 EINECS: 215-180-8 Merck Index: 14,4409 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...