Barium fluoride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36380-14
36380-22
CAAA36380-14EA
260.7
CAD
CAAA36380-14
CAAA36380-22
Barium fluoride ≥99.9% (metals basis)
Barium fluoride
MDL: MFCD00003450
Purity: 99.9%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: 3-6mm Fused Lump
Density: 4.83
Refractive Index: 1.4741
Odor: Odorless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Formula:
BaF₂ Boiling Pt: 2260 °C Melting Pt: 1353 °C Density: 4.83 Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS Number: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1,III Merck Index: 14,974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...