Cobalt ≥99.5% (metals basis excluding Ni), pieces ≤ 12 mm (0.47 in) for vacuum deposition (Ni ∼ 1%)

Supplier: Thermo Scientific Chemicals
Danger

36375-18 36375-30
CAAA36375-18EA 297.04 CAD
CAAA36375-18 CAAA36375-30
Cobalt ≥99.5% (metals basis excluding Ni), pieces ≤ 12 mm (0.47 in) for vacuum deposition (Ni ∼ 1%)
Cobalt

MDL: MFCD00010935


Purity: 99.5% CAS No.: 7440-48-4 Appearance: Gray Odor: Odorless MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR