Nitronium tetrafluoroborate ≥90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36374-06
36374-14
CAAA36374-06EA
100.8
CAD
CAAA36374-06
CAAA36374-14
Nitronium tetrafluoroborate ≥90%
Nitronium tetrafluoroborate
MDL: MFCD00011432
Fieser: 1,742 4,358 5,476 8,361 9,324 19,230
Purity: 90%
CAS No.: 13826-86-3
Molecular Formula: BF4NO2
Formula Weight: 132.81
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 240° dec.
UN No.: 3260
MDL No.: MFCD00011432
Formula:
BF₄NO₂ Melting Pt: 240 °C (dec.) Density: 1,26 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011432 CAS Number: 13826-86-3 EINECS: 237-533-5 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...