Erbium(III) oxide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

36372-14 36372-22
CAAA36372-14EA 301.78 CAD
CAAA36372-14 CAAA36372-22
Erbium(III) oxide ≥99.8% (metals basis)
Erbium (III) oxide

For vacuum deposition


Purity: 99.8% CAS No.: 12061-16-4 Molecular Formula: Er2O3 Formula Weight: 382.56 Physical Form: 3-12mm Sintered Lump Density: 8.64 Melting Point: 2400° Merck Reference: 14,3646 MDL No.: MFCD00010991


For vacuum deposition

Formula: Er₂O₃
Melting Pt: 2400 °C
Density: 8.64
Storage Temperature: Ambient
MDL Number: MFCD00010991
CAS Number: 12061-16-4
EINECS: 235-045-7
Merck Index: 14,3646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR