Dysprosium(III) oxide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Didysprosium trioxide, Dysprosium oxide

36370-14 36370-22
CAAA36370-14EA 312.84 CAD
CAAA36370-14 CAAA36370-22
Dysprosium(III) oxide ≥99.9% (REO, rare earth oxide basis)
Dysprosium (III) oxide

MDL: MFCD00010986 Notes: For vacuum deposition.


Purity: 99.9% CAS No.: 1308-87-8 Molecular Formula: Dy2O3 Formula Weight: 373.00 Physical Form: 3-12mm Sintered Lump Density: 7.81 Melting Point: 2340° Merck Reference: 14,3482 MDL No.: MFCD00010986

Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: 2400
Density: 7.81
Storage Temperature: Ambient
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR