Magnesium acetate tetrahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
36358-22
36358-36
36358-A4
CAAA36358-22EA
56.09
CAD
CAAA36358-22
CAAA36358-36
CAAA36358-A4
Magnesium acetate tetrahydrate 98.0-102.0% ACS
Magnesium acetate tetrahydrate
MDL: MFCD00149214
Beilstein Registry No.: 3730605
Purity: 98.0-102.0%
CAS No.: 16674-78-5
Molecular Formula: Mg(CH3COO)2.4H2O
Formula Weight: 214.45 (142.39anhy)
Physical Form: Crystalline
Density: 1.454
Melting Point: 80°
Beilstein Reference: 3730605
Merck Reference: 14,5654
MDL No.: MFCD00149214
Formula:
(H₃CCOO)₂Mg·4H₂O Melting Pt: 80 °C Density: 1.454 Storage Temperature: Ambient |
MDL Number:
MFCD00149214 CAS Number: 16674-78-5 EINECS: 205-554-9 Merck Index: 14,5654 |
Specification Test Results
Insoluble matter | 0.005% max. |
Cl | 0.001% max. |
SO3 | 0.005% max. |
Ba | 0.001% max. |
Ca | 0.01% max. |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm max. |
Mn | 0.001% max |
K | 0.005% max |
NA | 0.005% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...