Digadolinium trioxide ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Gadolinium (III) oxide
36355-14
36355-22
CAAA36355-14EA
216.46
CAD
CAAA36355-14
CAAA36355-22
Digadolinium trioxide ≥99.9% (REO, rare earth oxide basis)
Digadolinium trioxide
MDL: MFCD00011026
Notes: For vacuum deposition
Purity: 99.9%
CAS No.: 12064-62-9
Molecular Formula: Gd2O3
Formula Weight: 362.50
Physical Form: 3-12mm Sintered Lump
Density: 7.407
Melting Point: 2330°
Merck Reference: 14,4326
MDL No.: MFCD00011026
Formula:
Gd₂O₃ MW: 362.5 g/mol Melting Pt: 2330 °C Density: 7.407 Storage Temperature: Ambient |
MDL Number:
MFCD00011026 CAS Number: 12064-62-9 EINECS: 235-060-9 Merck Index: 14,4326 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...