Silicon(II) oxide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

36347-14 36347-22
CAAA36347-14EA 255.96 CAD
CAAA36347-14 CAAA36347-22
Silicon(II) oxide ≥99.99% (metals basis)
Silicon (II) oxide

MDL: MFCD00151536 Notes: For vacuum deposition


Purity: 99.99% CAS No.: 10097-28-6 Molecular Formula: OSi Formula Weight: 44.09 Physical Form: 3-6mm Lump, Outgassed Density: 2.13 Appearance: Black Odor: Odorless Melting Point: 1702° Boiling Point: 1880° Merck Reference: 14,8496 MDL No.: MFCD00151536

Formula: SiO
Boiling Pt: 1880 °C
Melting Pt: >1702 °C
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 14,8496

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR