Butyl stearate, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Stearic acid butyl ester, Butyl octadecanoate
36338-36
36338-A3
CAAA36338-36EA
103.81
CAD
CAAA36338-36
CAAA36338-A3
Butyl stearate, tech.
Butyl stearate
MDL: MFCD00026669
CAS No.: 123-95-5
Molecular Formula: CH3(CH2)16COO(CH2)3CH3
Formula Weight: 340.59
Flash Point: 160°(320°F)
Physical Form: Liquid
Density: 0.861
Refractive Index: 1.4430
Appearance: Colorless to pale yellow
Melting Point: 17-22°
Boiling Point: 343°
Beilstein Reference: 1792866
Merck Reference: 14,1589
MDL No.: MFCD00026669
Formula:
C₂₂H₄₄O₂ Boiling Pt: 343 °C Melting Pt: 17…22 °C Density: 0.861 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026669 CAS Number: 123-95-5 EINECS: 204-666-5 Merck Index: 14,1589 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...