Calcium carbonate ≥99.0% ACS low in alkali
Supplier: Thermo Scientific Chemicals
36337-30
36337-A1
CAAA36337-30EA
121.19
CAD
CAAA36337-30
CAAA36337-A1
Calcium carbonate ≥99.0% ACS low in alkali
Calcium carbonate
MDL: MFCD00010906
Purity: 99.0% min
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odor: Odorless
Melting Point: 800° dec.
Merck Reference: 14,1657
MDL No.: MFCD00010906
Formula:
CaCO₃ Melting Pt: 800 °C (dec.) Density: 2.930 Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |
Specification Test Results
Insoluble in dilute hydrochloric acid | 0.01% |
Cl | 0.1% |
F | 0.0015% |
SO3 | 0.005% |
Ba | 0.01% |
NH3 | 0.003% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.002% |
MG | 0.01% |
K | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...