Nickel (II) sulfate hexahydrate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickelous sulfate, Hexahydrate, Nickel sulfate hexahydrate, Nickel sulphate hexahydrate

36336-22 36336-36 36336-A4
CAAA36336-22EA 54.19 CAD
CAAA36336-22 CAAA36336-36 CAAA36336-A4
Nickel (II) sulfate hexahydrate ≥98.0% ACS
Nickel sulphate hexahydrate

MDL: MFCD00149813 Notes: m.p. 100°C -5H2O, 280°C -6H2O


Purity: 98.0% min CAS No.: 10101-97-0 Molecular Formula: NiO4S.6H2O Formula Weight: 262.86 (154.77anhy) Physical Form: Powder Density: 2.070 Appearance: Blue to bluish green Odor: Odorless pH: 4.3-4.7 Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00149813

Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840
Density: 2.070
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1 III
Merck Index: 14,6517

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.001%
Nitrogen compounds (as N) 0.002%
Co 0.002%
MG 0.005%
Mn 0.002%
K 0.0001
NA 0.0005
Cu 0.00005
Fe 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR