Aluminium fluoride, anhydrous ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36335-06
36335-14
36335-22
CAAA36335-06EA
152
CAD
CAAA36335-06
CAAA36335-14
CAAA36335-22
Aluminium fluoride, anhydrous ≥99.5%
Aluminium fluoride
For vacuum deposition
Purity: 99.5%
CAS No.: 7784-18-1
Molecular Formula: AlF3
Formula Weight: 83.98
Physical Form: 3-12mm Sintered Lump
Density: 2.882
Melting Point: 1291° subl.
Merck Reference: 14,339
MDL No.: MFCD00003426
For vacuum deposition
Formula:
AlF₃ Melting Pt: 1291 °C (subl.) Density: 2.882 Storage Temperature: Ambient |
MDL Number:
MFCD00003426 CAS Number: 7784-18-1 EINECS: 232-051-1 UN: 3260 ADR: 8,II Merck Index: 14,339 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...