Cadmium selenide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium (II) selenide

36323-06 36323-14 36323-22
CAAA36323-06EA 103.81 CAD
CAAA36323-06 CAAA36323-14 CAAA36323-22
Cadmium selenide ≥99.995% (metals basis)
Cadmium selenide

Vacuum deposition


Purity: 99.995% CAS No.: 1306-24-7 Molecular Formula: CdSe Formula Weight: 191.36 Physical Form: 3-12mm Sintered Lump Density: 5.81 Appearance: Brown-red to black Odor: Odorless Melting Point: 1350° Merck Reference: 14,1626 UN No.: 3077 MDL No.: MFCD00010917


Vacuum deposition

Formula: CdSe
MW: 191.37 g/mol
Melting Pt: 1268
Density: 5.81
Storage Temperature: Ambient
MDL Number: MFCD00010917
CAS Number: 1306-24-7
EINECS: 215-148-3
UN: 3283
ADR: 6.1 III
Merck Index: 14,1626

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR