Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zirconium dioxide, Zirconium Oxide
36319-18
36319-30
CAAA36319-18EA
170.64
CAD
CAAA36319-18
CAAA36319-30
Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)
Zirconium (IV) oxide
MDL: MFCD00011310
Notes: For vacuum deposition
Purity: 99.7%
CAS No.: 1314-23-4
Molecular Formula: ZrO2
Formula Weight: 123.22
Physical Form: 3-12mm Sintered Lump
Density: 5.89
Melting Point: ca 2700°
Boiling Point: ca 5000°
Merck Reference: 14,10180
MDL No.: MFCD00011310
Formula:
ZrO₂ MW: 123.22 g/mol Boiling Pt: ∼5000 °C Melting Pt: ∼2700 °C Density: 5.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...