Lead(II) telluride ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

36317-09 36317-18
CAAA36317-09EA 233.84 CAD
CAAA36317-09 CAAA36317-18
Lead(II) telluride ≥99.99% (metals basis)
Lead (II) telluride

In semiconductor research, photoconductor cells


Purity: 99.99% CAS No.: 1314-91-6 Molecular Formula: PbTe Formula Weight: 334.79 Physical Form: -325 Mesh Powder Density: 8.164 Melting Point: 917° Merck Reference: 14,5422 UN No.: 3284 MDL No.: MFCD00016274


In semiconductor research, photoconductor cells

Formula: PbTe
MW: 334.8
Melting Pt: 940
Density: 8.164
Storage Temperature: Ambient
MDL Number: MFCD00016274
CAS Number: 1314-91-6
EINECS: 215-247-1
UN: 3284
ADR: 6.1 III
Merck Index: 14,5422

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR