Manganese ≥99.95% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
36315-14
36315-22
CAAA36315-14EA
167.48
CAD
CAAA36315-14
CAAA36315-22
Manganese ≥99.95% (metals basis), powder -325 mesh
Manganese
MDL: MFCD00011111
Purity: 99.95%
CAS No.: 7439-96-5
UN No.: 3089
MDL No.: MFCD00011111
Formula:
Mn MW: 54.94 g/mol Density: 7.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...