Antimony(III) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Antimony trioxide, Diantimony trioxide, di-Antimony trioxide
36314-18
36314-30
CAAA36314-18EA
301.78
CAD
CAAA36314-18
CAAA36314-30
Antimony(III) oxide ≥99.9% (metals basis)
Antimony (III) oxide
MDL: MFCD00011214
Notes: For vacuum deposition
Purity: 99.9%
CAS No.: 1309-64-4
Molecular Formula: Sb2O3
Formula Weight: 291.50
Physical Form: 25mm and down Sintered Lump
Density: 5.67
Appearance: White
Odor: Odorless
Melting Point: 656°
Boiling Point: 1550° subl.
Merck Reference: 14,711
MDL No.: MFCD00011214
Formula:
O₃Sb₂ MW: 291.52 g/mol Boiling Pt: 1425 (subl.) Melting Pt: 654 Density: 5.67 Storage Temperature: Ambient |
MDL Number:
MFCD00011214 CAS Number: 1309-64-4 EINECS: 215-175-0 UN: 1549 ADR: 6.1 III Merck Index: 14,711 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...