Azelaic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36308-14
36308-22
36308-59
CAAA36308-14EA
64.62
CAD
CAAA36308-14
CAAA36308-22
CAAA36308-59
Azelaic acid 98%
Azelaic acid
MDL: MFCD00004432
Beilstein Registry No.: 1101094
Purity: 98%
CAS No.: 123-99-9
Molecular Formula: HOOC(CH2)7COOH
Formula Weight: 188.22
Flash Point: 215°(419°F)
Physical Form: Crystalline
Density: 1.225
Melting Point: ca 102°
Boiling Point: 286°/100mm
Beilstein Reference: 1101094
Merck Reference: 14,905
MDL No.: MFCD00004432
Formula:
C₉H₁₆O₄ Boiling Pt: 286 °C (100 mmHg) Melting Pt: ∼ 102 °C Density: 1.225 Flash Pt: 215 °C (419 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004432 CAS Number: 123-99-9 EINECS: 204-669-1 Merck Index: 14,905 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...