Tin(IV) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Tin(iv)oxide, Stannic Oxide, Tin dioxide
36302-14
36302-22
CAAA36302-14EA
199.08
CAD
CAAA36302-14
CAAA36302-22
Tin(IV) oxide ≥99.9% (metals basis)
Tin (IV) oxide
MDL: MFCD00011244
Notes: For vacuum deposition
Purity: 99.9%
CAS No.: 18282-10-5
Molecular Formula: O2Sn
Formula Weight: 150.69
Physical Form: 3-12mm Sintered Lump
Density: 6.95
Refractive Index: 1.9968
Appearance: White
Odor: Odorless
Melting Point: 1630°
Boiling Point: 1800-1900° subl.
Merck Reference: 14,8778
MDL No.: MFCD00011244
Formula:
O₂Sn MW: 150.71 g/mol Boiling Pt: 1800…1900 °C (subl.) Melting Pt: 1624.85 Density: 6.95 Storage Temperature: Ambient |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 EINECS: 242-159-0 Merck Index: 14,08778 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...