Nickel chromium alloy (80:20 w%) ≥99.9% (metals basis), lumps

Supplier: Thermo Scientific Chemicals
Warning

36298-14 36298-22
CAAA36298-14EA 237 CAD
CAAA36298-14 CAAA36298-22
Nickel chromium alloy (80:20 w%) ≥99.9% (metals basis), lumps
Nickel Chromium alloy (80:20 w%)

For vacuum deposition


Purity: 99.9% CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: 3-12mm Sintered Lump Odor: Odorless


For vacuum deposition

Storage Temperature: Ambient
MDL Number: MFCD00673110
CAS Number: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR