Aluminium sulfate hydrate 98.0-102% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum(III) sulfate xhydrate, Aluminum(III) sulphate xhydrate
36294-30
36294-A1
CAAA36294-30EA
63.83
CAD
CAAA36294-30
CAAA36294-A1
Aluminium sulfate hydrate 98.0-102% ACS
Aluminium sulphate hydrate
MDL: MFCD00149136
Purity: 98.0-102%
CAS No.: 17927-65-0
Molecular Formula: Al2(SO4)3.xH2O (x≈14-18)
Formula Weight: 342.15(anhy)
Physical Form: Crystalline
Density: 1.61
pH: 3.5
Merck Reference: 14,366
MDL No.: MFCD00149136
Formula:
Al₂O₁₂S₃·H₂O MW: 366.21 g/mol Density: 1.61 Storage Temperature: Ambient |
MDL Number:
MFCD00149136 CAS Number: 17927-65-0 EINECS: 233-135-0 Merck Index: 14,366 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.005% |
Ca | 0.01% |
MG | 0.002% |
K | 0.005% |
NA | 0.02% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.00002 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...