Aluminium, shot ACS 8-12 mm (0.31-0.47 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

36293-22 36293-36
CAAA36293-22EA 94.96 CAD
CAAA36293-22 CAAA36293-36
Aluminium, shot ACS 8-12 mm (0.31-0.47 in)
Aluminium

MDL: MFCD00134029 Notes: Flattened shot, typically 3-4mm thick


CAS No.: 7429-90-5 Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Specification Test Results

Matter insoluble in dilute hydrochloric acid 0.05% Max.
Si 0.1% Max.
Nitrogen compounds (as N) 0.001% Max.
Cu 0.02% Max.
Fe 0.1% Max.
Mn 0.002% Max.
Ti 0.03% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR