4-Amino-3-hydroxy-1-naphthalenesulfonic acid ≥90% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulfonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-naphthol-4-sulfonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulfonic acid, 1-Amino-2-hydroxy-4-naphthalenesulfonic acid
Purity: 90%
CAS No.: 116-63-2
Molecular Formula: C10H9NO4S
Formula Weight: 239.25
Physical Form: Powder
Melting Point: ca 295° dec.
Beilstein Reference: 2697469
Merck Reference: 14,454
MDL No.: MFCD00004019
Formula:
C₁₀H₉NO₄S MW: 239.25 g/mol Boiling Pt: 466 Melting Pt: ∼ 295 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004019 CAS Number: 116-63-2 EINECS: 204-147-3 Merck Index: 14,454 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...