Acetic acid glacial ≥99.7% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

36289-AE 36289-AP 36289-AY 36289-K3
CAAA36289-AEEA 88.32 CAD
CAAA36289-AE CAAA36289-AP CAAA36289-AY CAAA36289-K3
Acetic acid glacial ≥99.7% ACS
Acetic acid

MDL: MFCD00036152 Fieser: 2,5 5,3 7,1 8,1 19,1 21,1


Purity: 99.7+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odor: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Formula: H₃CCOOH
Boiling Pt: 118.1 °C
Melting Pt: 16.6 °C
Density: 1.049
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
Merck Index: 14,55
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Dilution Test Pass test
Evaporation residue 0.001%
(CH3CO)2O 0.01%
Cl 1 ppm
SO3 0.1%
Heavy Metals (as Pb) 0.5 ppm
Fe 0.2 ppm
Substances reducing dichromate Pass test
Substances reducing permanganate P.T. Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR