Antimony(III) telluride ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

36282-09 36282-18
CAAA36282-09EA 237 CAD
CAAA36282-09 CAAA36282-18
Antimony(III) telluride ≥99.999% (metals basis)
Antimony (III) telluride

For vacuum deposition


Purity: 99.999% CAS No.: 1327-50-0 Molecular Formula: Sb2Te3 Formula Weight: 626.30 Physical Form: 3-12mm Lump Density: 6.5 Appearance: Grey Odor: Odorless Melting Point: 629° UN No.: 1549 MDL No.: MFCD00016320


For vacuum deposition

Formula: Sb₂Te₃
MW: 626.32 g/mol
Melting Pt: 629 °C
Density: 6.5
Storage Temperature: Ambient
MDL Number: MFCD00016320
CAS Number: 1327-50-0
EINECS: 215-480-9
UN: 1549
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR