Antimony(III) chloride ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stripping tin coating from steel surfaces, in organic synthesis, as catalyst, in making other antimony salts, in chemical microscopy for drug identification
Purity: 99.0% min
CAS No.: 10025-91-9
Molecular Formula: SbCl3
Formula Weight: 228.11
Physical Form: Crystalline
Density: 3.14
Melting Point: 73.4°
Boiling Point: 223.5°
Solubility: Soluble in water with hydrolysis to SbOCl. Soluble in HCl solution, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone, carbon tetrachloride. Insoluble in organic bases
Merck Reference: 14,707
UN No.: 1733
MDL No.: MFCD00011212
Stripping tin coating from steel surfaces, in organic synthesis, as catalyst, in making other antimony salts, in chemical microscopy for drug identification
Formula:
SbCl₃ Boiling Pt: 223.5 °C Melting Pt: 73.4 °C Density: 3.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Merck Index: 14,707 |
Specification Test Results
Insoluble in chloroform | 0.05% |
SO3 | 0.005% |
As | 0.02% |
Ca | 0.005% |
Cu | 0.001% |
Fe | 0.002% |
PB | 0.005% |
K | 0.01% |
NA | 0.02%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...