Praseodymium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

36276-09 36276-18
CAAA36276-09EA 164.32 CAD
CAAA36276-09 CAAA36276-18
Praseodymium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis)
Praseodymium (III,IV) oxide

MDL: MFCD00011178 Notes: For vacuum deposition


Purity: 99.9% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: 3-12mm Sintered Lump Melting Point: 2500° MDL No.: MFCD00011178

Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2500 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR