Ethanolamine 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Aminoethanol, Monoethanolamine

36260-AP 36260-AY
CAAA36260-APEA 91.58 CAD
CAAA36260-AP CAAA36260-AY
Ethanolamine 99+% ACS
Ethanolamine

MDL: MFCD00008183 Beilstein Registry No.: 505944 Fieser: 1,357 2,189 4,222 16,162


Purity: 99+% CAS No.: 141-43-5 Molecular Formula: HOCH2CH2NH2 Formula Weight: 61.08 Flash Point: 93°(199°F) Physical Form: Liquid Density: 1.012 Refractive Index: 1.4540 Sensitivity: Air Sensitive and Hygroscopic Odor: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 171
Melting Pt: 10.5
Density: 1.012
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8 III
Merck Index: 14,3727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 15
H2O 0.3%
Fe 5ppm ppm
Heavy Metals 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR