Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
36259-03
36259-06
36259-14
CAAA36259-03EA
227.52
CAD
CAAA36259-03
CAAA36259-06
CAAA36259-14
Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS
Hexachloroplatinic (IV) acid hexahydrate
MDL: MFCD00149910
Fieser: 1890 487 13145 15135
Purity: 99.95%
CAS No.: 18497-13-7
Molecular Formula: H2Cl6Pt.6H2O
Formula Weight: 517.91 (409.82anhy)
Physical Form: Lump
Density: 2.430
Appearance: Red-brown
Odor: Odorless
Melting Point: 60° dec.
Merck Reference: 14,7526
UN No.: 2507
MDL No.: MFCD00149910
Formula:
H₂PtCl₆·6H₂O MW: 517.9 g/mol Melting Pt: 60 °C (dec.) Density: 2.430 Storage Temperature: Ambient |
MDL Number:
MFCD00149910 CAS Number: 18497-13-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III Merck Index: 14,7526 |
Specification Test Results
Solubility in alcohol | Pass Test |
Alkali and other salts (as sulfates) | 0.05% |
Suitability for potassium determinations | Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...