Zirconium ≥99.8% (metals basis excluding Hf), lumps 3-6 mm (0.1-0.2 in) (max. 4% Hf)

Supplier: Thermo Scientific Chemicals

36253-14 36253-22
CAAA36253-14EA 144.89 CAD
CAAA36253-14 CAAA36253-22
Zirconium ≥99.8% (metals basis excluding Hf), lumps 3-6 mm (0.1-0.2 in) (max. 4% Hf)
Zirconium

MDL: MFCD00011303


Purity: 99.8% CAS No.: 7440-67-7 Physical Form: For vacuum deposition Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR