Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
36252-14
36252-22
CAAA36252-14EA
295.46
CAD
CAAA36252-14
CAAA36252-22
Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis)
Ytterbium (III) oxide
MDL: MFCD00011290
Notes: For vacuum deposition.
Purity: 99.9%
CAS No.: 1314-37-0
Molecular Formula: Yb2O3
Formula Weight: 394.08
Physical Form: 3-12mm Sintered Lump
Density: 9.2
Melting Point: 2227°
Merck Reference: 14,10105
MDL No.: MFCD00011290
Formula:
O₃Yb₂ Melting Pt: 2227 °C Density: 9.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011290 CAS Number: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...