Resorcinol 99.0-100.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00002269
Beilstein Registry No.: 906905
Soluble in water, alcohol, ether, and glycerol
Purity: 99.0-100.5%
CAS No.: 108-46-3
Molecular Formula: C6H6O2
Formula Weight: 110.11
Flash Point: 171°(339°F)
Physical Form: Crystalline
Density: 1.272
Sensitivity: Air and Light Sensitive
Appearance: White to cream
Odor: Phenol-like
pH: 4-6
Melting Point: 109-112°
Boiling Point: 280-281°
Solubility: Soluble in water, alcohol, ether, and glycerol
Beilstein Reference: 906905
Merck Reference: 14,8155
UN No.: 2876
MDL No.: MFCD00002269
Formula:
C₆H₆O₂ Boiling Pt: 280…281 °C Melting Pt: 109…112 °C Density: 1.272 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002269 CAS Number: 108-46-3 EINECS: 203-585-2 UN: 2876 ADR: 6.1,III Merck Index: 14,8155 |
|
Specification Test Results
Form | Crystalline |
Insoluble matter | 0.005% |
Residue after ignition | 0.01% |
Titratable acid | 0.004meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...