Barium carbonate 99,0-101,0 ACS

Supplier: Thermo Scientific Chemicals
Warning

36236-36 36236-A1 36236-D3
CAAA36236-36EA 142.99 CAD
CAAA36236-36 CAAA36236-A1 CAAA36236-D3
Barium carbonate 99,0-101,0 ACS
Barium carbonate

MDL: MFCD00003448 Notes: m.p. 174°C (90 atm), 811°C (1 atm)


Purity: 99.0-101.0% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formula: BaCO₃
MW: 197.34 g/mol
Boiling Pt: Dec.
Melting Pt: 1350
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 14,969

Order Now

Specification Test Results

Insoluble in dilute hydrochloric acid 0.015%
Cl 0.2%
Water soluble titratable base 0.002meq/g
Oxidizing substances (as NO3) 0.005%
S 0.001%
K 0.005%
NA 0.02%
Ca 0.05%
Heavy Metals (as Pb) 0.001%
Fe 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR