Ammonium carbonate ≥30% (NH₃ basis) ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
CAS No.: 506-87-6
Molecular Formula: Approx. (NH4)2CO3
Formula Weight: 96.09
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Ammonia-like
Melting Point: 60° subl.
Solubility: Slowly soluble in water (1:4 portion). Decomposes in hot water
Beilstein Reference: 3627235
Merck Reference: 14,508
MDL No.: MFCD00010890
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
Formula:
(NH₄)₂CO₃ Melting Pt: 60 °C (subl.) Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010890 CAS Number: 506-87-6 EINECS: 208-058-0 Merck Index: 14,508 |
Specification Test Results
Insoluble matter | 0.005% |
Nonvolatile matter | 0.01% |
Cl | 5 ppm |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm max. |
Sulfur compounds (as SO3) | 0.002% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...