di-Ammonium oxalate monohydrate 99.0-101.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
Common Applications: In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals.
Purity: 99.0-101.0%
CAS No.: 6009-70-7
Molecular Formula: (COONH4)2.H2O
Formula Weight: 142.11 (124.10anhy)
Physical Form: Granular
Density: 1.50
Appearance: White
Melting Point: 70° dec.
Solubility: Soluble in water. Slightly soluble in alcohol
Beilstein Reference: 3759830
Merck Reference: 14,537
UN No.: 3288
MDL No.: MFCD00149694
In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
Formula:
(NH₄)₂C₂O₄·H₂O MW: 142.11 g/mol Boiling Pt: 70 °C (1013 hPa) Melting Pt: 70 °C (dec.) Density: 1.50 Storage Temperature: Ambient |
MDL Number:
MFCD00149694 CAS Number: 6009-70-7 EINECS: 214-202-3 UN: 3288 ADR: 6.1,III Merck Index: 14,537 |
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.02% |
Cl | 0.002% |
SO3 | 0.002% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 2% |
Color | White |
Form | Granular |
Solubility | Soluble in water, Slightly soluble in alcohol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...