Magnesium chloride hexahydrate 99.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
36226-36
36226-A3
CAAA36226-36EA
121.82
CAD
CAAA36226-36
CAAA36226-A3
Magnesium chloride hexahydrate 99.0-102.0% ACS
Magnesium chloride hexahydrate
MDL: MFCD00149781
Fieser: 10,251
Purity: 99.0-102.0%
CAS No.: 7791-18-6
Molecular Formula: MgCl2.6H2O
Formula Weight: 203.30 (95.22anhy)
Physical Form: Crystalline
Density: 1.570
Melting Point: 117° dec.
Merck Reference: 14,5662
MDL No.: MFCD00149781
Formula:
MgCl₂·6H₂O Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C (dec.) Density: 1.570 Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 14,5662 |
Specification Test Results
Insoluble matter | 0.005% |
NO3 | 0.001% |
PO3 | 5 ppm |
SO3 | 0.002% |
NH3 | 0.002% |
Ba | 0.005% |
Ca | 0.0001 |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm |
Mn | 5ppm% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...