Lithium carbonate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36225-36
36225-A3
CAAA36225-36EA
339.7
CAD
CAAA36225-36
CAAA36225-A3
Lithium carbonate ≥99.0% ACS
Lithium carbonate
MDL: MFCD00011084
Purity: 99.0% min
CAS No.: 554-13-2
Molecular Formula: Li2CO3
Formula Weight: 73.90
Physical Form: Powder
Density: 2.11
Appearance: White
Odor: Odorless
pH: 10-11.5
Melting Point: 723°
Boiling Point: 1310° dec.
Merck Reference: 14,5527
MDL No.: MFCD00011084
Formula:
Li₂CO₃ Boiling Pt: 1310 °C (dec.) Melting Pt: 723 °C Density: 2.11 Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 14,5527 |
Specification Test Results
Insoluble in dilute hydrochloric acid | 0.01% |
Cl | 0.005% |
NO5 | 5 ppm |
Sulfur compounds | 0.2% |
NH3 | 5 ppm max. |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.002% |
Ca | 0.01% |
K | 0.01%% |
NA | 0.1%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...