Diammonium hydrogen citrate 98.0-103.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36219-22
36219-36
36219-A3
CAAA36219-22EA
54.19
CAD
CAAA36219-22
CAAA36219-36
CAAA36219-A3
Diammonium hydrogen citrate 98.0-103.0% ACS
di-Ammonium hydrogen citrate
MDL: MFCD00013068
Purity: 98.0-103.0%
CAS No.: 3012-65-5
Molecular Formula: C6H14N2O7
Formula Weight: 226.19
Physical Form: Granular
Density: 1.48
Merck Reference: 14,512
MDL No.: MFCD00013068
Formula:
HOC(CO₂H)(CH₂CO₂NH₄)₂ Density: 1.48 Storage Temperature: Ambient |
MDL Number:
MFCD00013068 CAS Number: 3012-65-5 EINECS: 221-146-3 Merck Index: 14,512 |
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.01% |
Cl | 0.001% |
C2O3 | Pass test (limit about 0.05%) |
PO3 | 5 ppm max. |
SO3 | 0.005% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...