Lithium chloride ≥99% ACS

Supplier: Thermo Scientific Chemicals
Warning

36217-36 36217-A3
CAAA36217-36EA 336.54 CAD
CAAA36217-36 CAAA36217-A3
Lithium chloride ≥99% ACS
Lithium chloride

In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants


Purity: 99% min CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: Granular Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Solubility: Very soluble in water, alcohol, ether, pyridine, and nitrobenzene Merck Reference: 14,5528 MDL No.: MFCD00011078


In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now

Specification Test Results

Insoluble matter 0.01%
Titratable base 0.008meq/gmeq/g
Loss on drying at 105°C 1%
NO3 0.00001
SO3 0.01%
Ba 0.003%
Heavy Metals (as Pb) 0.002%
Fe 0.001%
Ca 0.01%%
K 0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR