Manganese ≥99.98% (metals basis), granules 0.8-12 mm (0.03-0.47 in)

Supplier: Thermo Scientific Chemicals

36215-14 36215-22
CAAA36215-14EA 148.84 CAD
CAAA36215-14 CAAA36215-22
Manganese ≥99.98% (metals basis), granules 0.8-12 mm (0.03-0.47 in)
Manganese

For vacuum deposition


Purity: 99.98% CAS No.: 7439-96-5 Appearance: Grey Odor: Odorless MDL No.: MFCD00011111


For vacuum deposition

Formula: Mn
MW: 54.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR