Magnesium oxide ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Magnesia usta
36214-14
36214-22
CAAA36214-14EA
165.9
CAD
CAAA36214-14
CAAA36214-22
Magnesium oxide ≥99.95% (metals basis)
Magnesium oxide
MDL: MFCD00011109
Notes: For vacuum deposition
Purity: 99.95%
CAS No.: 1309-48-4
Molecular Formula: MgO
Formula Weight: 40.30
Physical Form: 3-12mm Fused Lump
Density: 3.58
Refractive Index: 1.736
Odor: Odorless
Melting Point: 2852°
Boiling Point: 3600°
Merck Reference: 14,5677
MDL No.: MFCD00011109
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 14,5677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...