Ammonium metavanadate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

36213-18 36213-30
CAAA36213-18EA 82 CAD
CAAA36213-18 CAAA36213-30
Ammonium metavanadate ≥99.0% ACS
Ammonium metavanadate

MDL: MFCD00011430 Slightly soluble in water. Soluble in dilute ammonia


Purity: 99.0% min CAS No.: 7803-55-6 Molecular Formula: NH4VO3 Formula Weight: 116.98 Physical Form: Powder Density: 2.33 Melting Point: 200° dec. Solubility: Slightly soluble in water. Soluble in dilute ammonia Merck Reference: 14,568 UN No.: 2859 MDL No.: MFCD00011430

Formula: NH₄VO₃
Melting Pt: 200 °C (dec.)
Density: 2.33
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1,II
Merck Index: 14,568

Order Now

Specification Test Results

Solubility in ammonium hydroxide Pass test
CO3 Pass test (about 0.3%)
Cl 0.002
SO3 0.0005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR