Silicon ≥99.999% (metals basis), crystalline powder -100+325 mesh

Supplier: Thermo Scientific Chemicals
Warning

36212-09 36212-18 36212-30
CAAA36212-09EA 131.93 CAD
CAAA36212-09 CAAA36212-18 CAAA36212-30
Silicon ≥99.999% (metals basis), crystalline powder -100+325 mesh
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Appearance: White Odor: Odorless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR