Hafnium dioxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Supplier: Thermo Scientific Chemicals
36204-04
36204-09
36204-18
CAAA36204-04EA
59.88
CAD
CAAA36204-04
CAAA36204-09
CAAA36204-18
Hafnium dioxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Hafnium dioxide
MDL: MFCD00003565
Notes: For vacuum deposition, Monoclinic
Purity: 99.9%
CAS No.: 12055-23-1
Molecular Formula: HfO2
Formula Weight: 210.49
Physical Form: 3-12mm Sintered Lump
Density: 9.68
Melting Point: 2774°
Merck Reference: 14,4588
MDL No.: MFCD00003565
Formula:
HfO₂ MW: 210.49 Boiling Pt: 5400 Melting Pt: 2812 Density: 9.68 Storage Temperature: Ambient |
MDL Number:
MFCD00003565 CAS Number: 12055-23-1 EINECS: 235-013-2 Merck Index: 14,04727 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...