Titanium(IV) oxide anatase ≥99.6% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pigment White 6, Titanium dioxide anatase, Anatase

36199-14 36199-22 36199-36
CAAA36199-14EA 66.99 CAD
CAAA36199-14 CAAA36199-22 CAAA36199-36
Titanium(IV) oxide anatase ≥99.6% (metals basis)
Titanium (IV) oxide anatase

MDL: MFCD00011269


Purity: 99.6% CAS No.: 1317-70-0 Molecular Formula: O2Ti Formula Weight: 79.90 Physical Form: -325 Mesh Powder Density: 3.9 Merck Reference: 14,9472 MDL No.: MFCD00011269

Formula: TiO₂
MW: 79.88 g/mol
Density: 3.9
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 1317-70-0
EINECS: 215-280-1
Merck Index: 14,9472

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR